Principaux documents
BAF199170155
Scienceware® replacement hydrophobic filters
for use with vacuum aspirator collection system
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.79
Conditionnement
pack of 12 ea
Fabricant/nom de marque
Bel-Art F19917-0155
Description générale
Replaceable hydrophobic filter protects the pump motor from fluid contamination.
Informations légales
Scienceware is a registered trademark of SP Scienceware, a division of SP