Principaux documents
BAB617003300
SP Bel-Art® Durac® 4-Channel Electronic Timer and Clock
with certificate of calibration
About This Item
Code UNSPSC :
41112303
Nomenclature NACRES :
NB.01
Conditionnement
pack of 1 ea
Fabricant/nom de marque
HB Instruments B61700-3300
Plage de mesure
accuracy: ±3 sec, resolution: 0.01
H × l
80 mm (3.1 in.) × 80 mm (3.1 in.)
Description générale
This 4-channel timer has a capacity of 99 hours, 59 minutes and 59 seconds or can be switched to a 12- or 24-hour clock.
Caractéristiques et avantages
  • Counts either up or down; remembers the last time set
  • Lay flat on a bench, use the flip-open stand for a vertical view, or carry it on a lab coat with the convenient pocket clip
  • Requires one AAA battery (included)
  • Features a Certificate of Calibration and individual serial number, these electronic timers are tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Durac is a registered trademark of SP Scienceware, a division of SP