BAB614007100
SP Bel-Art® H-B Beaker Clip Liquid-in-Glass Thermometer Holder
stainless steel, holds 4 thermometers
Synonyme(s) :
thermometer holder
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.79
Produits recommandés
Matériaux
stainless steel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
HB Instruments B61400-7100
Largeur × L
38 mm (1.4 in.) × 3 in. (76 mm)
Description générale
Holds up to 4 thermometer probes with 3.5-4.0mm O.D. at one time.
Caractéristiques et avantages
Holds up to 4 thermometer probes with 3.5-4.0mm O.D. at one time.
- Clips easily onto the side of any beaker
- Made from SUS304 stainless steel
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP