SRP0284
Sphingosine kinase 1a Active from mouse
recombinant, expressed in baculovirus, ≥70% (SDS-PAGE)
Synonym(s):
SK1, SPHK1, SPK 1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352204
NACRES:
NA.54
Recommended Products
recombinant
expressed in baculovirus
Assay
≥70% (SDS-PAGE)
form
aqueous solution
specific activity
≥20 units/μg
mol wt
47.1 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
mouse ... SPHK1(8877)
General description
Mouse Sphingosine kinase 1 (GenBank Accession No. AF415213), full length with N-terminal His tag, MW = 47.1 kDa, expressed in a Baculovirus infected Sf9 cell expression system.
Application
Sphingosine kinase 1a is useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling. Sphingosine kinase 1a has been used in a study to assess the design of sphingosine kinase inhibitors to promote apoptosis of chemotherapeutic resistant cancer cells. Sphingosine kinase 1a has also been used in a study to investigate novel biomarkers for early prostate cancer detection.
Unit Definition
One unit of sphingosine kinase activity is defined as the amount of enzyme required to produce 1 pmol of S1P/min
Physical form
Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 50% glycerol and 3 mM DTT.
Preparation Note
Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Keng Gat Lim et al.
The international journal of biochemistry & cell biology, 44(9), 1457-1464 (2012-05-29)
Sphingosine kinase 1 catalyses the formation of the bioactive lipid, sphingosine 1-phosphate and is a target for anti-cancer agents. We demonstrate here that 2-(p-hydroxyanilino)-4-(p-chlorophenyl)thiazole (SKi, also referred to as SKI-II), FTY720 (Fingolimod), and (S)-FTY720 vinylphosphonate inhibit sphingosine kinase 1 activity
J Nunes et al.
British journal of cancer, 106(5), 909-915 (2012-02-09)
Current markers available for screening normal populations and for monitoring prostate cancer (PCa) treatment lack sensitivity and selectivity. Sphingosine-1-phosphate (S1P) is a circulating lipid second messenger involved in cell growth and migration, the immune response, angiogenesis, and malignant transformation. Eighty-eight
Brian M Barth et al.
Journal of neuroscience research, 90(1), 229-242 (2011-09-21)
Inflammation accompanied by severe oxidative stress plays a vital role in the orchestration and progression of neurodegeneration prevalent in chronic and acute central nervous system pathologies as well as in aging. The proinflammatory cytokine tumor necrosis factor-α (TNFα) elicits the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service