Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3725

Sigma-Aldrich

Icotinib

≥98% (HPLC)

Synonym(s):

(3-Ethynyl-phenyl)(7,8,10,11,13,14-hexahydro-6,9,12,15-tetraoxa-1,3-diazacyclododeca[b]naphthalen-4-yl)-amine, 4-[(3-Ethynylphenyl)amino]-6,7-benzo-12-crown-4-quinazoline, BPI 2009, BPI 2009H free base, BPI-2009, BPI-2009H free base, BPI2009, BPI2009H free base, Conmana

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21N3O4
CAS Number:
Molecular Weight:
391.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(c4cc(ccc4)C#C)c1ncnc2c1cc3c(c2)OCCOCCOCCO3

InChI key

QQLKULDARVNMAL-UHFFFAOYSA-N

Biochem/physiol Actions

Icotinib is an orally active, potent and highly selective epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor (IC50 = 5 nM) that inhibits EGFR-mediated intracellular tyrosine phosphorylation (IC50 = 45 nM; A431 cells) and cancer proliferation in cultures (A431/BGC-823 IC50 = 1.0/4.1 μM) and in vivo (30-120 mg/kg via daily p.o. to mice with A431, A549, H460 and HCT8 xenografts).
Orally active, potent and highly selective epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor with anti-cancer efficacy in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service