Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB3700864

Sigma-Aldrich

Anti-Rabbit IgG (Fc specific)-Alkaline Phosphatase antibody produced in donkey

affinity isolated antibody, buffered aqueous solution

Synonym(s):

ALP, Alk Phos

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

donkey

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rabbit

concentration

1.0 mg/mL

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Rabbit IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Alkaline Phosphatase (calf intestine), Anti-Donkey Serum, Rabbit IgG, Rabbit IgG F(c) and Rabbit Serum. No reaction was observed against Rabbit IgG F(ab′)2.

Immunogen

Rabbit IgG F(c) fragment

Physical properties

Antibody format: IgG

Physical form

Supplied in 0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0 with 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasushi Sawanobori et al.
PloS one, 9(10), e109995-e109995 (2014-10-22)
Thymic epithelial cells (TECs) are thought to play an essential role in T cell development and have been detected mainly in mice using lectin binding and antibodies to keratins. Our aim in the present study was to create a precise
Michela Bolla et al.
International archives of allergy and immunology, 164(2), 112-121 (2014-06-20)
Pomegranate allergy is associated with sensitization to non-specific lipid transfer proteins (nsLTPs). Our aim was to identify and characterize the non-specific nsLTPs expressed in pomegranate at the molecular level and to study their allergenic properties in terms of immunoglobulin E
Jullyana S S Quintans et al.
Molecules (Basel, Switzerland), 19(6), 8303-8316 (2014-06-21)
Hecogenin is a steroidal sapogenin largely drawn from the plants of the genus Agave, commonly known as 'sisal', and is one of the important precursors used by the pharmaceutical industry for the synthesis of steroid hormones. Hecogenin acetate (HA) is
Roberta Russo et al.
The FEBS journal, 281(17), 3828-3843 (2014-07-22)
Growing evidence suggests that the transcription factors belonging to the Jun family are involved in many important cellular events, such as the control of bone development in mammalians. We have characterized, for the first time, a member of the Jun

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service