MilliporeSigma
  • Pomegranate ( Punica granatum L.) expresses several nsLTP isoforms characterized by different immunoglobulin E-binding properties.
Pomegranate ( Punica granatum L.) expresses several nsLTP isoforms characterized by different immunoglobulin E-binding properties.
International archives of allergy and immunology (2014-06-20)
Michela Bolla, Sara Zenoni, Stephan Scheurer, Stefan Vieths, Maria Del Mar San Miguel Moncin, Mario Olivieri, Andrea Antico, Marta Ferrer, Felicia Berroa, Ernesto Enrique, Linda Avesani, Francesco Marsano, Gianni Zoccatelli
ABSTRACT
Pomegranate allergy is associated with sensitization to non-specific lipid transfer proteins (nsLTPs). Our aim was to identify and characterize the non-specific nsLTPs expressed in pomegranate at the molecular level and to study their allergenic properties in terms of immunoglobulin E (IgE)-binding and cross-reactivity with peach nsLTP (Pru p 3). A non-equilibrium two-dimensional (2-D) electrophoretic approach based on acid-urea PAGE and sodium dodecyl sulfate PAGE was set up to separate pomegranate nsLTPs. Their immunoreactivity was tested by immunoblotting carried out with anti-Pru p 3 polyclonal antibodies and sera from pomegranate-allergic patients. For final identification, pomegranate nsLTPs were purified by chromatography and subjected to trypsin digestion and mass spectrometry (MS) analysis. For this purpose, the sequences obtained by cDNA cloning of three pomegranate nsLTPs were integrated in the database that was subsequently searched for MS data interpretation. Four nsLTPs were identified by 2-D immunoblotting. The detected proteins showed different IgE-binding capacity and partial cross-reactivity with Pru p 3. cDNA cloning and MS analyses led to the identification of three nsLTP isoforms with 66-68% amino acid sequence identity named Pun g 1.0101, Pun g 1.0201 and Pun g 1.0301. By 2-D electrophoresis, we could separate different nsLTP isoforms possessing different IgE-binding properties, which might reflect peculiar allergenic potencies. The contribution of Pru p 3 to prime sensitization is not central as in other plant nsLTPs.
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, ≥99%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
SAFC
Iodoacetamide
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg