Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

S0438

Sigma-Aldrich

Anti-Sodium Channel PN4 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Sodium Channel Scn8a

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot (chemiluminescent): 1:200

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCN8A(6334)
mouse ... Scn8a(20273)
rat ... Scn8a(29710)

Immunogen

synthetic peptide corresponding to residues 1042-1061 of mouse or rat Scn8a (intracellular epitope). The peptide is highly homologous (19/20 residues identical) in human.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tzoumaka, E. et al.
The Journal of Neuroscience, 60, 37-37 (2000)
Ligia A Papale et al.
Human molecular genetics, 18(9), 1633-1641 (2009-03-04)
In a chemical mutagenesis screen, we identified the novel Scn8a(8J) allele of the gene encoding the neuronal voltage-gated sodium channel Na(v)1.6. The missense mutation V929F in this allele alters an evolutionarily conserved residue in the pore loop of domain 2
Youngsoon Kim et al.
PloS one, 7(3), e33664-e33664 (2012-03-23)
Myelinating Schwann cells (mSCs) form myelin in the peripheral nervous system. Because of the works by us and others, matrix metalloproteinase-9 (MMP-9) has recently emerged as an essential component of the Schwann cell signaling network during sciatic nerve regeneration. In
Lisa M Sharkey et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(9), 2733-2741 (2009-03-06)
The ENU-induced neurological mutant ataxia3 was mapped to distal mouse chromosome 15. Sequencing of the positional candidate gene Scn8a encoding the sodium channel Na(v)1.6 identified a T>C transition in exon 1 resulting in the amino acid substitution p.S21P near the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service