MilliporeSigma
  • Heterozygous mutations of the voltage-gated sodium channel SCN8A are associated with spike-wave discharges and absence epilepsy in mice.
Heterozygous mutations of the voltage-gated sodium channel SCN8A are associated with spike-wave discharges and absence epilepsy in mice.
Human molecular genetics (2009-03-04)
Ligia A Papale, Barbara Beyer, Julie M Jones, Lisa M Sharkey, Sergio Tufik, Michael Epstein, Verity A Letts, Miriam H Meisler, Wayne N Frankel, Andrew Escayg
ABSTRACT
In a chemical mutagenesis screen, we identified the novel Scn8a(8J) allele of the gene encoding the neuronal voltage-gated sodium channel Na(v)1.6. The missense mutation V929F in this allele alters an evolutionarily conserved residue in the pore loop of domain 2 of Na(v)1.6. Electroencephalography (EEG) revealed well-defined spike-wave discharges (SWD), the hallmark of absence epilepsy, in Scn8a(8J) heterozygotes and in heterozygotes for two classical Scn8a alleles, Scn8a(med) (null) and Scn8a(med-jo) (missense). Mouse strain background had a significant effect on SWD, with mutants on the C3HeB/FeJ strain showing a higher incidence than on C57BL/6J. The abnormal EEG patterns in heterozygous mutant mice and the influence of genetic background on SWD make SCN8A an attractive candidate gene for common human absence epilepsy, a genetically complex disorder.