Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA017270

Sigma-Aldrich

Anti-ST3GAL2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Alpha 2,3-ST, Anti-Beta-galactoside alpha-2,3-sialyltransferase, Anti-CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase, Anti-Gal-NAc6S, Anti-Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase, Anti-SIAT4-B, Anti-ST3Gal II, Anti-ST3GalA2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

ATLPYLDSGALDGTHRVKLVPGYAGLQRLSKERLSGKSCACRRCMGDAGASDWFDSHFDGNISPVWTRENMDLPPDVQRWWMMLQPQFKSHNTNEVLEKLFQIVPGENPYRFRDPHQCRRCAVVGNSGNLRGSGYGQDVD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ST3GAL2(6483)

General description

ST3GAL2 (ST3 β-galactoside α-2,3-sialyltransferase 2) is a type II transmembrane protein consisting of two putative N-glycosylation sites (Asn(92) and Asn(211)). It is localized along the proximal compartments of the Golgi complex. It exerts enzymatic activity. It shows tissue-specific expression majorly in the skeletal muscle and heart. It is also expressed in the lung and kidney at a very low level.

Immunogen

CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ST3GAL2 (ST3 β-galactoside α-2,3-sialyltransferase 2) is mainly involved in the GD1a and GT1b ganglioside biosynthesis in the brain. It has been suggested that the ST3GAL2 expression depends on the promoter activity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72085

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y J Kim et al.
Biochemical and biophysical research communications, 228(2), 324-327 (1996-11-12)
A cDNA of human Gal beta 1,3GalNAc alpha 2,3-sialytransferase (hST3Gal II) which has been known to exhibit much more acceptor substrate preference for glycolipid than for O-linked oligosaccharides of glycoproteins, was isolated from the human liver cDNA library by plaque
Akiyoshi Taniguchi et al.
Biochemical and biophysical research communications, 300(2), 570-576 (2002-12-31)
In this report, we describe transcriptional regulation of the human Gal beta 1,3 GalNAc alpha 2,3-sialyltransferase II (hST3Gal II) gene. The results of 5'-RACE showed that the forms of two mRNAs differed only in the 5'-untranslated region (Types 1 and
Fernando M Ruggiero et al.
The Biochemical journal, 469(1), 83-95 (2015-04-29)
ST3Gal-II, a type II transmembrane protein, is the main mammalian sialyltransferase responsible for GD1a and GT1b ganglioside biosynthesis in brain. It contains two putative N-glycosylation sites (Asn(92) and Asn(211)). Whereas Asn(92) is only conserved in mammalian species, Asn(211) is highly conserved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service