Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4702018

Monoclonal Anti-CRP purified produced in mouse

new
Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

unconjugated

Quality Level

antibody product type

primary antibodies

clone

C2, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/ml

technique(s)

ELISA: suitable
immunohistochemistry: suitable

isotype

IgG1

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CRP(1401)

General description

The C-reactive protein (CRP) is a cyclic pentameric pentraxin family acute phase protein composed of five identical noncovalently bound nonglycosylated subunits (each subunit 24 kDa; physiologic CRP molecule 117,5 kDa). CRP is produced by the liver and its plasma levels rise dramatically during inflammatory processes occuring in the body. CRP is an initiator of classical complement cascade, binds to several nuclear components (chromatin, histones, etc.) and is also believed to play an important role in innate immunity. Patients with elevated basal levels of CRP are at increased risk for hypertension and cardiovascular disease.

Specificity

The mouse monoclonal antibody C2 recognizes human CRP, an 117 kDa plasma acute phase protein; it recognizes the antigen both in the presence and in the absence (samples containing EDTA) of Ca2+.

Immunogen

Purified human CRP

Physical form

Phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified by protein-A affinity chromatography.

Storage and Stability

Store at 2-8°C. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service