Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

HPA006344

Sigma-Aldrich

Anti-AKAP12 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-A-kinase anchor protein 12 antibody produced in rabbit, Anti-A-kinase anchor protein 250 kDa antibody produced in rabbit, Anti-AKAP 250 antibody produced in rabbit, Anti-Myasthenia gravis autoantigen gravin antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500
western blot: 0.04-0.4 μg/mL

immunogen sequence

QRAEAERPEEQAEASGLKKETDVVLKVDAQEAKTEPFTQGKVVGQTTPESFEKAPQVTESIESSELVTTCQAETLAGVKSQEMVMEQAIPPDSVETPTDSETDGST

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AKAP12(9590)

General description

AKAP12 (A-kinase anchoring protein 12) is a scaffold protein, and is also called gravin in humans. It has a wide range of tissue expression. Its subcellular distribution depends upon its isoform, which are three in number- α, β and γ. It has a molecular weight of 300kDa.

Immunogen

A-kinase anchor protein 12 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

AKAP12 (A-kinase anchoring protein 12) interacts with the RII subunit of protein kinase A (PKA), and also interacts with PKC, phosphodiesterase 4D, Ca2+/calmodulin, and the β2-adrenergic receptor. Thus, it has multiple essential roles in cellular processes. It sequesters Src protein to caveolin-rich lipid rafts, and thus, prevents the interaction of Src with FAK-associated adhesion and signaling complexes. Therefore, it inhibits the movement of tumor cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86532

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Yildirim et al.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology, 18(3), 635-640 (2013-09-26)
: Colorectal cancer (CRC) is a common and potentially lethal disease. A number of genetic aberrations is known to take place in colorectal carcinogenesis, which leads to progressive alteration of normal mechanisms controlling cell growth. A-kinase-anchoring protein 12 (AKAP12) plays
Micah B Schott et al.
Cellular signalling, 25(11), 2125-2135 (2013-07-11)
A-Kinase Anchoring Proteins (AKAPs) direct the flow of cellular information by positioning multiprotein signaling complexes into proximity with effector proteins. However, certain AKAPs are not stationary but can undergo spatiotemporal redistribution in response to stimuli. Gravin, a 300kD AKAP that
Benjamin Goeppert et al.
Journal of neuropathology and experimental neurology, 72(10), 933-941 (2013-09-18)
The scaffold protein A-kinase anchor protein 12 (AKAP12) exerts tumor suppressor activity and is downregulated in several tumor entities. We characterized AKAP12 expression and regulation in astrocytomas, including pilocytic and diffusely infiltrating astrocytomas. We examined 194 human gliomas and 23
B Su et al.
Oncogene, 32(16), 2016-2026 (2012-06-20)
Metastatic cell migration and invasion are regulated by altered adhesion-mediated signaling to the actin-based cytoskeleton via activated Src-FAK complexes. Src-suppressed C-kinase substrate (SSeCKS, the rodent orthologue of human Gravin/AKAP12), whose expression is downregulated by oncogenic Src and in many human
Nicholas W Bateman et al.
Journal of proteome research, 14(4), 1900-1910 (2015-03-10)
A majority of high-grade (HG) serous ovarian cancer (SOC) patients develop resistant disease despite high initial response rates to platinum/paclitaxel-based chemotherapy. We identified shed/secreted proteins in preclinical models of paclitaxel-resistant human HGSOC models and correlated these candidate proteins with patient

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service