EP0030081234
Eppendorf® BioBased ep Dualfilter T.I.P.S. SealMax
volume 0.5—20 μL, sterile, Biopur, L -46 mm, pkg of 960 ea (10 trays x 96 tips)
About This Item
RNase free
graduations
Biopur
Recommended Products
sterility
sterile
feature
DNase free
RNase free
graduations
Biopur
packaging
pkg of 960 ea (10 trays x 96 tips)
manufacturer/tradename
Eppendorf® 0030081234
greener alternative product characteristics
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
DNA purification: suitable
PCR: suitable
L
-46 mm
volume
0.5—20 μL
suitability
suitable for PCR
suitable for microbiology
suitable for molecular biology
suitable for nucleic acid purification
greener alternative category
General description
Application
Features and Benefits
- Improve your laboratory carbonfootprint and switch to epT.I.P.S. BioBased in the newEppendorf Reload variants. These pipette tips are produced from second-generationbiobased polymer with 90% of the feedstock attributed to renewable resources (i.e. waste and residues from vegetable oilrefining or waste cooking oil), avoiding the use of fossil fuel-based petroleum
- Reduce your laboratory waste volume by useing Reloads instead of disposable Racks
- Eppendorf production facility is ISCC PLUS(International Sustainability & Carbon Certification) certified, confirmingthe raw materials used at the facility are “bio-circular”. Enables traceabilityalong the entire supply chain
- Pipette tips with ACT (Accountability, Consistency, and Transparency) Label- an independent environmental impact factor assessment through non-profit MyGreen Lab
- Material, quality and purity certified
- Get excellent sample integrity as no slip agents, plasticizers and biocides are used
- Independent laboratory – Lot-specific purity certificates can be downloaded from Eppendorf website
- General quality certificates in compliance with standards and general Eppendorf quality items
- Available in volumes from 10 µL to 1,250 µL Long in Reloads for use with epT.I.P.S. Box 2.0
- Available as epT.I.P.S. BioBased Biopur, ep Dualfilter T.I.P.S. BioBased PCR clean and sterile, ep Dualfilter T.I.P.S. SealMax BioBased Biopur
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)