EP0030081064
Eppendorf® BioBased ep Dualfilter T.I.P.S.
volume 2—100 μL, sterile, PCR clean, L -53 mm, pkg of 960 ea (10 trays x 96 tips)
About This Item
PCR clean
RNase free
graduations
Recommended Products
sterility
sterile
feature
DNase free
PCR clean
RNase free
graduations
packaging
pkg of 960 ea (10 trays x 96 tips)
manufacturer/tradename
Eppendorf® 0030081064
greener alternative product characteristics
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
DNA purification: suitable
PCR: suitable
L
-53 mm
volume
2—100 μL
suitability
suitable for PCR
suitable for microbiology
suitable for molecular biology
suitable for nucleic acid purification
greener alternative category
General description
Application
Features and Benefits
- Improve your laboratory carbon footprint and switch to epT.I.P.S. BioBased in the new Eppendorf Reload variants. These pipette tips are produced from second-generation biobased polymer with 90% of the feedstock attributed to renewable resources (i.e. waste and residues from vegetable oilrefining or waste cooking oil), avoiding the use of fossil fuel-based petroleum.
- Reduce your laboratory waste volume – use Reloads instead of disposable Racks.
- Eppendorf production facility is ISCC PLUS(International Sustainability & Carbon Certification) certified, confirmingthe raw materials used at the facility are “bio-circular”. Enables traceabilityalong the entire supply chain.Pipette tips with ACT (Accountability, Consistency,and Transparency) Label - an independent environmental impact factor assessmentthrough non-profit My Green.
- Material that is both quality and purity certified.
- Get excellent sample integrity as no slip agents, plasticizers and biocides are used Independent laboratory – Lot-specific purity certificates can be downloaded from Eppendorf website.
- General quality certificates in compliance with standards and general Eppendorf quality items.
- Available in volumes from 10 µL to 1,250 µL Long in Reloads for use with epT.I.P.S. Box 2.0.
- Available as epT.I.P.S. BioBased Biopur, ep Dualfilter T.I.P.S. BioBased PCR clean and sterile, ep Dualfilter T.I.P.S. SealMax BioBased Biopur
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)