BMSIPD9600BD4
Ball Dropper to Fill 96 BeadBlaster™ 96 Well Plate
diam. 4 mm
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NA.82
Recommended Products
material
acrylic
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPD9600-BD4
diam.
4 mm
Related Categories
General description
This ball dropper is used to easily fill 96 well plates with 1 bead per well.
Application
Homogenization/milling/grinding of samples
Legal Information
BeadBlaster is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)