Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE251

Sigma-Aldrich

Anti-N-CoR Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Nuclear receptor corepressor 1, N-CoR, N-CoR1, Retinoid X receptor-interacting protein 13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NCOR1(9611)

General description

In the absence of ligands, the corepressor N-CoR represses nuclear hormone receptor-mediated gene transcription. In the presence of ligands, these hormone receptors activate gene expression and play critical roles in development, growth, and homeostasis. N-Cor-mediated transcriptional repression is facilitated through the formation of histone deacetylase (HDACs) N-CoR complexes or by conjugation from small ubiquitin-like modifier 1 (SUMO-1). It is shown that the SANT1 domain of N-CoR serves as a binding region for SUMO-E2 conjugating enzyme Ubc9 and SUMO-E3 ligase Pias1, which is necessary for N-CoR-mediated repression of respective reporter genes. SUMOylation-deficient N-COR knockout mice display a signification reduction in the repression of gene transcription and result in defects involving the differentiation and development of neural cells into erythrocytes and thymocytes.

Immunogen

KLH-conjugated linear peptide corresponding to a region between amino acids 1510 and 1540 of the N-CoR protein.

Application

Anti-N-CoR Antibody is a Rabbit Polyclonal Antibody for detection of N-CoR also known as Nuclear receptor corepressor 1 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Hormones & Receptors

Quality

Evaluated by Western Blot in RAW 264.7 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected N-CoR in RAW 264.7 cell lysate. No tween for best results.

Target description

~270 kDa observed. Uniprot describes 2 isoforms produced by alternative splicing at MW of 266 kDa and 270 kDa

Linkage

Replaces: 06-892

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
RAW 264.7 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sunmi Seok et al.
The Journal of clinical investigation, 128(7), 3144-3159 (2018-06-19)
Jumonji D3 (JMJD3) histone demethylase epigenetically regulates development and differentiation, immunity, and tumorigenesis by demethylating a gene repression histone mark, H3K27-me3, but a role for JMJD3 in metabolic regulation has not been described. SIRT1 deacetylase maintains energy balance during fasting
Giuseppina Mastrototaro et al.
Frontiers in cell and developmental biology, 9, 641410-641410 (2021-03-13)
TBL1XR1 gene is associated with multiple developmental disorders presenting several neurological aspects. The relative protein is involved in the modulation of important cellular pathways and master regulators of transcriptional output, including nuclear receptor repressors, Wnt signaling, and MECP2 protein. However
Juu-Chin Lu et al.
PloS one, 8(8), e71517-e71517 (2013-08-21)
In obesity, high levels of tumor necrosis factor α (TNFα) stimulate lipolysis in adipocytes, leading to hyperlipidemia and insulin resistance. Thiazolidinediones (TZDs), the insulin-sensitizing drugs, antagonize TNFα-induced lipolysis in adipocytes, thereby increasing insulin sensitivity in diabetes patients. The cellular target
Yu Jin Jung et al.
Stem cell research, 44, 101771-101771 (2020-03-29)
SIRT1 (NAD+-dependent deacetylase) plays a suppressive role during the late stages of adipogenesis. However, the effects of SIRT1 on the early phases of adipogenic differentiation from embryonic stem cells (ESCs) are poorly understood. We employed Sirt1+/+ and Sirt1-/- mouse embryonic
Zhiwei Luo et al.
Cell regeneration (London, England), 8(1), 21-29 (2019-06-18)
Class IIa histone deacetylases (HDACs) are a subfamily of HDACs with important functions in development and adult tissue homeostasis. As opposed to other HDACs, they lack catalytic function and bind transcription factors to recruit transcriptional co-regulators, mostly co-repressors such as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service