Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

233105

Sigma-Aldrich

CL-82198

A selective inhibitor of MMP-13 (IC₅₀ = 10 µM).

Synonym(s):

CL-82198

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22N2O3 · xHCl
CAS Number:
Molecular Weight:
302.37 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

light yellow

solubility

water: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Cl.N3(CCOCC3)CCCCNC(=O)c1[o]c2c(c1)cccc2

InChI

1S/C17H22N2O3.ClH/c20-17(16-13-14-5-1-2-6-15(14)22-16)18-7-3-4-8-19-9-11-21-12-10-19;/h1-2,5-6,13H,3-4,7-12H2,(H,18,20);1H

InChI key

PIOACXKZWXHBRB-UHFFFAOYSA-N

General description

A selective inhibitor of MMP-13 (IC50 = 10 µM) that does not appear to act by chelating Zn2+. Binds within the entire S1′ pocket of MMP-13, docking with the morpholine ring adjacent to the catalytic zinc atom. Does not inhibit MMP-1, MMP-9, and TACE.

Biochem/physiol Actions

Cell permeable: no
Primary Target
MMP-13
Product does not compete with ATP.
Reversible: no
Target IC50: 10 µM against MMP-13

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze at -20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chen, J.M., et al. 2000. J. Am. Chem. Soc.122, 9648.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service