Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

939749

Sigma-Aldrich

Sulfonated poly(ether ether ketone)

new

proton conducting polymer, degree of sulfonation 70%

Synonym(s):

Polyaromatic sulfonic acid, Sulfonated PEEK, Sulfonated aromatic polymer, Sulfonated poly(arylene ether sulfone) ionomer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C19H12O6S)n(C19H12O3)m
UNSPSC Code:
13111024

Quality Level

form

powder, crystals or chunks

mol wt

100-200 kDa

color

white to beige-brown

functional group

sulfonic acid

polymer architecture

shape: linear

General description

Sulfonated poly(ether ether ketone) (sulfonated PEEK) is a specialized polymeric material derived from the modification of polyetheretherketone (PEEK). By incorporating sulfonic acid groups into its structure, we obtain a versatile, high-performance powder with a molecular weight of 150 kDa and a degree of sulfonation of 70%. This alteration introduces new properties and enhances its performance in specific applications, making sulfonated PEEK an attractive material for a wide range of uses. Its unique characteristics include excellent thermal and chemical stability, high proton conductivity, and resistance to degradation under harsh conditions.

Application

The exceptional properties of sulfonated PEEK make it an ideal choice for various applications, most notably in fuel cell technology and water purification. In fuel cells, sulfonated PEEK functions as a proton exchange membrane (PEM), allowing the transport of protons while preventing the mixing of gases. Its ability to operate at high temperatures and maintain proton conductivity contributes to the overall efficiency of fuel cell systems. Additionally, sulfonated PEEK′s high proton conductivity is leveraged in water purification and desalination processes, facilitating the separation of ions and impurities for effective water treatment. Beyond these applications, sulfonated PEEK is employed in high-performance capacitors, sensors, and various industries such as aerospace, automotive, and electronics, where its thermal resistance, chemical durability, and proton conductivity are essential for reliable performance in demanding environments.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service