Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.04703

Supelco

Chromate standard solution certified reference material 1000 mg/L CrO4

new

certified reference material, 1000 mg/L (Cl), traceable to SRM of NIST K₂CrO₄ in H₂O, Certipur®

Sign Into View Organizational & Contract Pricing


About This Item

grade

certified reference material

Quality Level

description

technique(s): ion chromatography: suitable

product line

Certipur®

form

liquid

concentration

1000 mg/L (Cl)

density

1.0 g/cm3 at 20 °C

format

single component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST. Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation. Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date

Packaging

500ml in HD-PE bottle

Legal Information

Certipur is a registered trademark of Merck KGaA, Darmstadt, Germany.
CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service