1.04688
Fluoride standard solution

certified reference material, 1000 mg/L (F), traceable to SRM from NIST NaF in H₂O, Certipur®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
certified reference material
Quality Level
description
(technique(s): ion chromatography: suitable)
product line
Certipur®
form
liquid
concentration
1000 mg/L (F)
density
1.000 g/cm3 at 20 °C
format
single component solution
storage temp.
15-25°C
General description
This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST. Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation. Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Packaging
500ml in HD-PE bottle
Legal Information
Certipur is a registered trademark of Merck KGaA, Darmstadt, Germany.
CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service