Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

938718

Sigma-Aldrich

Pd(COD)(DQ)

greener alternative

>95%

Synonym(s):

(1,5-Cyclooctadiene)(duroquinone) palladium(0), Pd(COD)(DQ) (COD=1,5-cyclooctadiene,DQ=duroquinone)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O2Pd
CAS Number:
Molecular Weight:
378.80
UNSPSC Code:
12352100

Quality Level

Assay

>95%

form

powder or crystals

reaction suitability

reagent type: catalyst
core: palladium
reaction type: Buchwald-Hartwig Amination

reagent type: catalyst
core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: catalyst
core: palladium
reaction type: Cross Couplings

reagent type: catalyst
core: palladium
reaction type: Heck Reaction

reagent type: catalyst
core: palladium
reaction type: Hiyama Coupling

reagent type: catalyst
core: palladium
reaction type: Miyaura Borylation Reaction

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

light brown to dark brown

greener alternative category

storage temp.

2-8°C

SMILES string

O=C(C(C)=C(C1=O)C)C(C)=C1C.C[Pd](C)(C)C.C2C/C=C\CC/C=C\2

InChI

InChI=1S/C10H12O2.C8H12.Pd/c1-5-6(2)10(12)8(4)7(3)9(5)11;1-2-4-6-8-7-5-3-1;/h1-4H3;1-2,7-8H,3-6H2;/b;2-1-,8-7-;

InChI key

LDTNOVDKFHVJGF-GHDUESPLSA-N

General description

Pd(COD)(DQ) is a rare example of a robust, air-stable, phosphine free 18 electron Pd(0) complex. The complex easily undergoes facile ligand exchange with a range of ancilliary ligands to give useful palladium compounds suitable for a range of catalytic transformations.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Zero-valent palladium species are active catalytic species in a range of useful catalytic reactions, including C-C coupling, C-N coupling, allylic substituion and alkene funcitonilizaiton reactions.

Features and Benefits

Pd(COD)(DQ) offers:

  • High thermal and air stability.
  • Benign ligands that do not interfere in catalytic reactions once ligand exchange has occurred.
  • Good solution stability.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service