Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

937428

Sigma-Aldrich

Rockphos Pd G6 Br

greener alternative

Synonym(s):

2,3-dihydro-1H-indole-2-thione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H53BrF3OPPd
Molecular Weight:
800.12
UNSPSC Code:
12352142

form

powder or crystals

Quality Level

reaction suitability

reaction type: Cross Couplings

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

light green to dark green-beige

functional group

(Palladium)
phosphine

greener alternative category

storage temp.

2-8°C

SMILES string

CC1=C(C2=C(C(C)C)C=C(C(C)C)C=C2C(C)C)C([P]([Pd](Br)C3=CC=C(C(F)(F)F)C=C3)(C(C)(C)C)C(C)(C)C)=C(OC)C=C1

General description

Rockphos Pd G6 Br is a Gen 6 Buchwald precatalyst. The G6 precatalysts are air-stable, palladium-based oxidative addition complexes (OACs) that offer an alternative to the previously developed classes of palladacycle precatalysts. Unlike previous generations, the bulkiest biarylphosphine ligand precatalysts are readily prepared. These precatalysts have been shown to be effective for many reactions, including C-N, C-O, and C-F cross couplings.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service