Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

746266

Sigma-Aldrich

Oxamyl-1-13C

99 atom % 13C, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC6H13N3O3S
Molecular Weight:
220.25
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

95% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+1

SMILES string

CN(C)C(/[13C](SC)=N/OC(NC)=O)=O

InChI

1S/C7H13N3O3S/c1-8-7(12)13-9-5(14-4)6(11)10(2)3/h1-4H3,(H,8,12)/b9-5-/i5+1

InChI key

KZAUOCCYDRDERY-BDPMGFTRSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service