Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

768545

Sigma-Aldrich

Ethyl-d5 chloroformate solution

7 wt. % in toluene, 99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3D5O2Cl
Molecular Weight:
113.55
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % D

form

solution

concentration

7 wt. % in toluene

mass shift

M+5

storage temp.

2-8°C

SMILES string

ClC(OC([2H])([2H])C([2H])([2H])[2H])=O

InChI

1S/C3H5ClO2/c1-2-6-3(4)5/h2H2,1H3/i1D3,2D2

InChI key

RIFGWPKJUGCATF-ZBJDZAJPSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service