Skip to Content
Merck
All Photos(1)

Key Documents

PHL23762

Monascin

new

phyproof® Reference Substance

Synonym(s):

(3S,3aR,9aR)-3-Hexanoyl-9a-methyl-6-((E)-propenyl)-3a,4,8,9a-tetrahydro-3H-furo[3,2-g]isochromene-2,9-dione, (3S,3aR,9aR)-3a,4,8,9a-Tetrahydro-9a-methyl-3-(1-oxohexyl)-6-(1E)-1-propenyl-2H-furo[3,2-g][2]benzopyran-2,9(3H)-dione, Monascoflavin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O5
CAS Number:
Molecular Weight:
358.43
Beilstein:
51355
MDL number:
UNSPSC Code:
41116107

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

technique(s)

HPLC: suitable

application(s)

food and beverages
pharmaceutical

format

neat

storage temp.

−20°C

SMILES string

CCCCCC(=O)[C@@H]1[C@H]2CC3=C(COC(\C=C\C)=C3)C(=O)[C@]2(C)OC1=O

InChI

1S/C21H26O5/c1-4-6-7-9-17(22)18-16-11-13-10-14(8-5-2)25-12-15(13)19(23)21(16,3)26-20(18)24/h5,8,10,16,18H,4,6-7,9,11-12H2,1-3H3/b8-5+/t16-,18+,21-/m1/s1

InChI key

XXKNHBAFFJINCK-RVEJDSBJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Monascin is one of the azaphilonoid pigments in extracts of Monascus pilosus-fermented rice (red-mold rice). It is a potent inhibitor of carcinogenesis measured against chemical or UV initiated, phorbol-promoted mouse skin tumors.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service