
Monascin

Synonym(s):
(3S,3aR,9aR)-3-Hexanoyl-9a-methyl-6-((E)-propenyl)-3a,4,8,9a-tetrahydro-3H-furo[3,2-g]isochromene-2,9-dione, (3S,3aR,9aR)-3a,4,8,9a-Tetrahydro-9a-methyl-3-(1-oxohexyl)-6-(1E)-1-propenyl-2H-furo[3,2-g][2]benzopyran-2,9(3H)-dione, Monascoflavin
Empirical Formula (Hill Notation):
C21H26O5
CAS Number:
Molecular Weight:
358.43
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing