Skip to Content
Merck
All Photos(1)

Key Documents

TOU (TOU I-2)

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human blood

packaging

tube of 5 μg 93093001-DNA-5UG
pkg of vial of cells 93093001-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human B lymphocyte, EBV transformed

Cell Line Description

Peripheral blood lymphocytes from a male Tunisian immortalised by EBV transformation. The cell line contains the following deletions, in the human immunoglobulin heavy chain constant region locus (IGH chromosome 14q32.3): deletion II (del EP1-A1-GP) Gm-Am genotype Gm b; -; -A2m2/Gm g; za; .A2m2. Gm allotypes are indicated in the order G3m; G1m; G2m. The absence of G1m and G2m allotypes due to the deletion of the IGHG1 and IGHG2 genes is indicated by (-). Only one allotype is defined on the gamma 2 chain and may or may not be present; (.) indicates the absence of this allotype.

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures at 5-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service