MOH (TOU II-3)
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
human blood
packaging
tube of 5 μg 93093002-DNA-5UG
pkg of vial of cells 93093002-1VL
growth mode
Suspension
karyotype
Not specified
morphology
Lymphoblast
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human B lymphocyte, EBV transformed
Cell Line Description
Peripheral blood lymphocytes from a male Tunisian immortalised by EBV transformation. The cell line contains the following deletions in the human immunoglobulin heavy chain constant region locus: deletion I (G1-EP1-A1-GP-G2-G4) and deletion II (EP1-A1-GP). Gm-Am phenotype: Gmb:-;A2m2/Gmg;za;.A2m2. The absence of G2m allotypes due to the deletion of the IGHG2 gene.
Application
Molecular and immunological analysis of gene deletions
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service