GF79315027
Titanium
wire reel, 5m, diameter 0.25mm, annealed, 99.6+%
Synonym(s):
Titanium, TI005122
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
MDL number:
UNSPSC Code:
12141746
PubChem Substance ID:
NACRES:
NA.23
Assay
99.6%
form
wire
autoignition temp.
860 °F
manufacturer/tradename
Goodfellow 793-150-27
resistivity
42.0 μΩ-cm, 20°C
L × diam.
5 m × 0.25 mm
bp
3287 °C (lit.)
mp
1660 °C (lit.)
density
4.5 g/mL at 25 °C (lit.)
SMILES string
[Ti]
InChI
1S/Ti
InChI key
RTAQQCXQSZGOHL-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Wei-Qiang Yu et al.
Journal of nanoscience and nanotechnology, 14(6), 4387-4393 (2014-04-18)
The TiO2 nanotubes by anodization have been extensively studied for medical implant and orthopedic applications because of enhancing bone development. In the present study, a new nano-foveolae structure verified by SEM and AFM was prepared by simulating the nanotubes exfoliation
Shaoming Yang et al.
Journal of nanoscience and nanotechnology, 14(6), 4292-4296 (2014-04-18)
The structural and functional characterizations of titanium nitride (TiN) advanced ceramic microsprings (CMSs), with a coil diameter of several micrometers and synthesized by chemical vapor deposition (CVD) were investigated by microscopy techniques. The CMSs were sufficiently mechanically elastic for extension
Bijuan Zheng et al.
Journal of nanoscience and nanotechnology, 14(5), 3527-3531 (2014-04-17)
Pd-modified N-doped TiO2 nanoparticles were prepared by the sol-gel method. The X-ray diffraction (XRD) pattern indicated that the pure anatase TiO2 has been obtained. Transmission electron microscope (TEM) was used to observe the micro-morphology of the nanoparticles. The average size
Liming Wang et al.
Journal of nanoscience and nanotechnology, 14(6), 4224-4228 (2014-04-18)
Tetragonal Barium titanate (BaTiO3) nanotube arrays have been prepared using the template-assisted hydrothermal method combined with an annealing process. The in-situ chemical conversion of TiO2 nanotube array templates ensured that BaTiO3 maintained the morphology of the nanotube architectures. Moreover, X-ray
Konstantinos X Michalakis et al.
The Journal of oral implantology, 40(2), 146-152 (2014-05-02)
Dental implants with an internal connection have been designed to establish a better stress distribution when lateral external forces act on the prosthesis and minimize the forces transmitted to the fastening screw. In the present study, 10 externally and 10
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service