Skip to Content
Merck

124249

Sigma-Aldrich

4-tert-Butylcatechol

97%

Sign Into View Organizational & Contract Pricing

About This Item

Linear Formula:
(CH3)3CC6H3-1,2-(OH)2
CAS Number:
Molecular Weight:
166.22
Beilstein:
2043335
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Assay

97%

bp

285 °C (lit.)

mp

52-55 °C (lit.)

SMILES string

CC(C)(C)c1ccc(O)c(O)c1

InChI

1S/C10H14O2/c1-10(2,3)7-4-5-8(11)9(12)6-7/h4-6,11-12H,1-3H3

InChI key

XESZUVZBAMCAEJ-UHFFFAOYSA-N

General description

4-tert-Butylcatechol undergoes electrochemical trimerization via anodic oxidation and mechanism has been studied by cyclic voltammetry and controlled-potential coulometry. It inhibits the activity of tyrosinase at concentrations higher than 1×10−3M. It undergoes oxidation with laccase to yield quinones which on reaction with dienes and oxidation afford naphthoquinones.

Application

4-tert-Butylcatechol was used in the synthesis of tungsten oxide nanoparticles by nonaqueous sol-gel process.


Still not finding the right product?

Explore all of our products under 4-tert-Butylcatechol


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library