1.02375
N,N-Dimethylformamide
Synonym(s):
DMF, N,N-Dimethylmethanamide, Formic acid dimethylamide, DMF, NSC 5356
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
MDL number:
UNSPSC Code:
12352111
Bp:
153 °C (lit.)
Vapor pressure:
2.7 mmHg ( 20 °C)
vapor density
2.5 (vs air)
vapor pressure
2.7 mmHg ( 20 °C)
form
liquid
autoignition temp.
833 °F
expl. lim.
15.2 %
refractive index
n20/D 1.430 (lit.)
pH
7 (20 °C, 200 g/L)
bp
153 °C (lit.)
mp
−61 °C (lit.)
density
0.944 g/mL (lit.)
SMILES string
[H]C(=O)N(C)C
InChI
1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3
InChI key
ZMXDDKWLCZADIW-UHFFFAOYSA-N
Application
Solvent for many hydrophobic organic compounds.
Still not finding the right product?
Explore all of our products under N,N-Dimethylformamide
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.