501743
Carbon Monoxide (7%), Carbon Dioxide (15%), Oxygen (4%), and Methane (4.5%).
In in nitrogen, cylinder of 14 L, analytical standard
About This Item
Recommended Products
grade
analytical standard
form
gas
packaging
cylinder of 14 L
manufacturer/tradename
Scott Gas T237-14
availability
not available in Canada, EU
concentration
In in nitrogen
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
format
multi-component solution
Looking for similar products? Visit Product Comparison Guide
Application
Analyte
Application
regulator
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Inhalation - Repr. 1A - STOT RE 2 Inhalation
Storage Class
2A - Gases
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service