SPR-488B
StressMarq Biosciences Inc
StressMarq Amyloid Beta 1-42 Oligomers
100 µg
Synonym(s):
Abeta Oligomers, Abeta peptide, Amyloid beta peptide oligomers, APP
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Molecular Weight:
4.5
Recommended Products
assay
95
concentration
2 mg/mL
application(s)
life science and biopharma
biological source
human
mol wt
predicted mol wt 4.5 kDa
pH
7.4
storage temp.
-80 °C
storage condition
avoid repeated freeze/thaw cycles
manufacturer/tradename
StressMarq Biosciences Inc
Related Categories
General description
Human Synthetic Amyloid Beta 1-42 Oligomers are certified >95% pure using mass spec and HPLC. They have been determined to be low endotoxin <2.5 EU/mL @ 1mg/mL. Validated for WB | In vivo Assay | In vitro Assay. Monomer source is catalog# SPR-485.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.