Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ZRB2913

Sigma-Aldrich

Anti-TRF2 Antibody, clone 1N14 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

TTAGGG repeat-binding factor 2, Telomeric DNA-binding protein, Telomeric repeat-binding factor 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1N14, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 59.59 kDa
observed mol wt ~65 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgGκ

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... TERF2(7014)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1N14 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects Telomeric repeat-binding factor 2/TRF2. It targets an epitope within 24 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the N-terminal region of human Telomeric repeat-binding factor 2.

Application

Quality Control Testing

Evaluated by Western Blotting in Daudi cell lysate.

Western Blotting Analysis (WB): A 1:10,000 dilution of this antibody detected TRF2 in lysates from Daudi cells.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected TRF2 in nuclear extract of U-2 OS cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected TRF2 in human tonsil tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TRF2 in U-2 OS cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Telomeric repeat-binding factor 2 (UniProt: Q15554; also known as TTAGGG repeat-binding factor 2, Telomeric DNA-binding protein, TERF2) is encoded by the TERF2 (also known as TRBF2, TRF2) gene (Gene ID: 7014) in human. TERF2 is homodimeric protein that is ubiquitous in its expression. Higher expression levels are reported in spleen, thymus, prostate, uterus, testis, peripheral blood leukocytes, colon, and small intestine. Two soforms of TERF2 are reported that are produced by alternative splicing. TRF2, like TRF1, contains a Myb-related DNA binding motif. TERF2 binds to telomeric double-stranded 5′-TTAGGG-3′ repeat and plays a central role in telomere maintenance and protection against end-to-end fusion of chromosomes. It is also required to recruit a number of factors and enzymes required for telomere protection, including the sheltering complex, TERF2IP/RAP1 and DCLRE1B/Apollo. TEFR2 colocalizes with telomeric DNA in interphase cells and is located at chromosome ends during metaphase. It is phosphorylated upon DNA damage and the phosphorylated TERF2 is not bound to telomeric DNA, and rapidly localizes to damage sites. Inhibition of TRF2 causes apoptosis in several mammalian cell types and its over-expression is observed in a variety of human cancers. As, TRF2 is a critical component of the telomere protection machinery, with its dysregulation contributing to various pathologies, particularly those related to chromosomal instability and cellular aging. Its roles in telomere maintenance, protection, and length regulation make it a significant focus in the study of aging and cancer biology.This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service