Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

ZRB2724

Sigma-Aldrich

Anti-CD86 Antibody, clone 2H2 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Activation B7-2 antigen, B70, BU63, CD28LG2, CTLA-4 counter-receptor B7.2, FUN-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2H2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 38 kDa
observed mol wt ~55 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgGκ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... CD86(942)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2H2 is a ZooMAb® rabbit recombinant monoclonal antibody that detects CD86. It targets an epitope within 16 amino acids from the extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the extracellular domain of human CD86.

Application

Quality Control Testing

Evaluated by Western Blotting with MCF-7.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected CD86 in MCF-7 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected CD86 in human spleen tissue lysate and a 1:50,000 dilution detected T7/HIS-tagged, human recombinant CD86.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected CD86 in human tonsil tissue sections.

Flow Cytometry Analysis: 1 µg from a representative lot detected CD86 in one million human peripheral blood mononuclear cells (PBMC).

Affinity Binding Assay: A representative lot of this antibody bound CD86 peptide with at least one thousand-fold (1,000X) higher than with non-specific control peptide.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CD86 in HDLM-2 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

T-lymphocyte activation antigen CD86 (UniProt: P42081; also known as Activation B7-2 antigen, B70, BU63, CTLA-4 counter-receptor B7.2, FUN-1) is encoded by the CD86 (also known as CD28LG2) gene (Gene ID: 942) in human. CD86 is a type I transmembrane protein belonging to the immunoglobulin superfamily, containing an extracellular Ig-like V-type (aa 33-131) and Ig-like C2-type (aa 15-225) domains essential for T cell interaction, a transmembrane domain (aa 248-268) and a cytoplasmic domain. The extracellular domain of CD86 is responsible for binding to CD28 or CTLA-4 on T cells. CD86 is expressed on the surface of antigen-presenting cells, including dendritic cells, macrophages, and B cells. Its expression is upregulated upon activation of these cells, which typically occurs during an immune response to pathogens. Its dysregulation has been implicated in autoimmune diseases such as rheumatoid arthritis and multiple sclerosis, allergic reactions, and transplant rejection, making it a potential target for immunotherapy. In some cancers, the manipulation of CD86 and its pathways is explored for therapeutic cancer vaccines.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service