Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

ZRB2685

Sigma-Aldrich

Anti-SOX5 Antibody, clone 1I9 ZooMAb® Rabbit Monoclonal

enhanced validation

recombinant, expressed in HEK 293 cells

Synonym(s):

SOX-5, Transcription factor SOX-5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1I9, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 84.03 kDa
observed mol wt ~84 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

rat

packaging

antibody small pack of 25

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... SOX5(6660)

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1I9 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Transcription factor SOX-5. It targets an epitope within 24 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the C-terminal region of human Transcription factor SOX-5.

Application

Quality Control Testing

Evaluated by Western Blotting with DDK-tagged, full-length, Human SOX5.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected DDK-tagged, full-length, recombinant Human SOX5.

Tested Applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected SOX5 in MCF7 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected SOX5 in Human prostate tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound SOX5 with a KD of 4.5 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Transcription factor SOX-5 (UniProt: P35711; also known as SOX5) is encoded by the SOX5 gene (Gene ID: 6660) in human. SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. SOX5 is a transcription factor that is involved in in chondrocytes differentiation and cartilage formation. It is reported to specifically bind the 5′-AACAAT-3′ DNA motif present in enhancers and super-enhancers and promotes expression of genes important for chondrogenesis, including cartilage matrix protein-coding genes, such as COL2A1 and AGC1. It is required for overt chondrogenesis when condensed pre-chondrocytes differentiate into early-stage chondrocytes. Together with SOX6, it maintains a pool of highly proliferating chondroblasts between epiphyses and metaphyses, to form columnar chondroblasts and delay chondrocyte pre-hypertrophy. SOX-5 has also been linked to the migration and invasion of various cancer cells, including hepatocellular carcinoma, melanoma, and prostate cancer. It is also shown to promote the migration and invasion of fibroblast-like synoviocytes in part by regulating MMP-9 expression in collagen-induced arthritis. Mutations in SOX5 gene have been linked to Lamb-Shaffer syndrome, an autosomal dominant, neurodevelopmental disorder that is characterized by developmental delay, intellectual disability, and motor impairment. Five isoforms of SOX5 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Shi, Y., et al. (2018). Front. Immunol. 9; 749; Wang, D., et al. (2015). Med. Oncol. 32(2); 461).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service