Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

ZRB2549

Anti-CDC20 Antibody, clone 2C9 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Cell division cycle protein 20 homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2C9, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 54.72 kDa
observed mol wt ~58 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

porcine, rat, mouse

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgGκ

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... CDC20(991)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2C9 is a ZooMAb® rabbit recombinant monoclonal antibody that detects CDC20. It targets an epitope within 22 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the internal region of human CDC20.

Application

Quality Control Testing

Evaluated by Western Blotting in human testis tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CDC20 in human testis tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CDC20 in human tonsil tissue lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected CDC20 in human tonsil tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CDC20 in A-431 cells.

Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected CDC20 in human tonsil tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound with CDC20 peptide with at least one million-fold (1,000,000X) higher than with non-specific control peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Cell division cycle protein 20 homolog (UniProt: Q12834; also known as Cdc20, p55CDC) is encoded by the CDC20 gene (Gene ID: 991) in human. CDC20 contains an N-terminal region with a C-box, KEN-box, Mad2 interacting motif (MIM), and CRY-box motif, and a C-terminal region with seven WD40 repeats and an Ile-Arg (IR) motif. CDC20 plays a critical role regulating cell cycle progression and apoptosis. It acts as an activator of the anaphase-promoting complex (APC) and forms the APCCdc20 E3 ubiquitin ligase complex that is responsible for tagging specific proteins for degradation, thus facilitating mitosis. The APCCdc20 ubiquitin signaling pathway involved in other biological processes as well, including dendritic morphogenesis, brain development and plasticity in the mammalian brain. Cdc20 overexpression is associated with tumorigenesis and poor prognosis. Its downregulation is associated with DNA damage, impaired repair process, and apoptosis. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: He, W. and Meng, J., (2023), Am J Transl Res., 15(2):678 693.; Kim, A. H., et al., (2009), Cell 136(2), 322-36.; Neuert, H., et al., (2017), Development, 144(24), 4673-4683.)

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service