Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

ZRB2490

Sigma-Aldrich

Anti-CD5L Antibody, clone 2D23 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Apoptosis inhibitor expressed by macrophages, CD5 antigen-like, CT-2, IgM-associated peptide, SP-alpha, hAIM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2D23, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 38.09 kDa
observed mol wt ~28 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... CD5L(922)

Related Categories

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2D23 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects CD5L. It targets an epitope within 25 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the N-terminal half of human CD5L.

Application

Quality Control Testing

Evaluated by Western Blotting with His/ABP-tagged recombinant fragment of Human CD5L.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected His/ABP-tagged recombinant fragment of Human CD5L.

Tested Applications

Enzyme Immunoassay (ELISA) Analysis: A serial of dilutions from a representative lot detected His/ABP-tagged recombinant fragment of Human CD5L.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected CD5L in Human spleen tissue sections.

Affinity Binding Assay:: A representative lot of this antibody bound His/ABP-tagged recombinant fragment of Human CD5L with a KD of 2.7 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

CD5 antigen-like (UniProt: O43866; also known as Apoptosis inhibitor expressed by macrophages, hAIM, CT-2, IgM-associated peptide, SP-alpha) is encoded by the CD5L (also known as AP16, Q203/PRO229) gene (Gene ID: 922) in human. CD5L is a secreted glycoprotein of the SRCR superfamily that is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off to produce the mature form. Its expression is transcriptionally controlled by LXRs and its expression is observed in macrophages, spleen, lymph node, thymus, bone marrow, and fetal tissue. However, it is not detected in non-lymphoid tissues. It circulates in serum in relatively high amounts and serves as a serum biomarker for inflammatory conditions. It is involved in a broad spectrum of biological functions. It participates in the pathogenesis of inflammatory processes by preventing the apoptosis of macrophages and other cell types. It also acts as a key regulator of lipid synthesis. CD5L is reported to reduce lipid droplet size in adipocytes. Following incorporation into mature adipocytes via CD36-mediated endocytosis, it associates with cytosolic fatty acid synthase activity and inhibits its activity and leads to lipolysis, degradation of triacylglycerols into glycerol and free fatty acids. It participates in obesity-associated inflammation following recruitment of inflammatory macrophages into adipose tissues, a cause of insulin resistance and obesity-related metabolic disorders. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Sanjurjo, L., et al. (2018). Front. Immunol. 9; 480; Sanjurjo, L.et al. (2015). J. Leukocyte Biol. 98(2); 173-184).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service