Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

ZRB2451

Sigma-Aldrich

Anti-PROKR2 Antibody, clone 1J6 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-ATP5A1 Antibody, clone 1J7 ZooMAb® Rabbit Monoclonal, G-protein coupled receptor 73-like 1, G-protein coupled receptor I5E, GPR73b, GPRg2, PK-R2, PKR2, Prokineticin receptor 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1J6, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 44 kDa
observed mol wt ~42 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... PROKR2(128674)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1J6 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Prokineticin receptor 2 (PROKR2). It targets an epitope within 16 amino acids from the first extracellular domain in the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal, first extracellular domain of human Prokineticin receptor 2 (PROKR2).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell membrane extract.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PROKR2 in HeLa cell membrane extract.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PROKR2 in A431 and HeLa cell lysates.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PROKR2 in THP-1 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected PROKR2 in Human cerebral cortex tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Prokineticin receptor 2 (UniProt: Q8NFJ6; also known as PK-R2, PKR2, G-protein coupled receptor 73-like 1, G-protein coupled receptor I5E, GPR73b, GPRg2) is encoded by the PROKR2 (also known as GPR73L1, PKR2) gene (Gene ID: 128674) in human. Prokineticins bind to two different G protein-coupled receptors known as prokineticin receptor-1 and -2 (PROKR1 and PROKR2) that display about 85% sequence identity. They bind to PROK1 and PROK2 with nanomolar range affinities. PROKR2 is a multi-pass membrane protein with seven transmembrane domains. Its expression is observed in the ileocecum, thyroid gland, pituitary gland, salivary gland, adrenal gland, testis, ovary, and the brain. It is exclusively coupled to the G(q) subclass of heteromeric G proteins. Its activation leads to mobilization of calcium, stimulation of phosphoinositide turnover, and activation of p44/p42 mitogen-activated protein kinase. It is involved in normal development of the olfactory bulb and sexual maturation. Mice lackin

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service