ZRB2416
Anti-SOCS-3 Antibody, clone 2N6 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
CIS-3, Cytokine-inducible SH2 protein 3, SSI-3, STAT-induced STAT inhibitor 3, Suppressor of cytokine signaling 3
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
2N6, recombinant monoclonal
description
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 24.77 kDa
observed mol wt ~28 kDa
purified by
using Protein A
species reactivity
human
species reactivity (predicted by homology)
rat, mouse
packaging
antibody small pack of 25
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
technique(s)
affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
C-terminal
Protein ID accession no.
UniProt accession no.
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... SOCS3(9021)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in A375 cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SOCS-3 in A375 cell lysate.
Tested Applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected SOCS-3 in lysate from Human liver tissue and His/ABP-tagged recombinant fragment of Human SOCS-3.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected SOCS-3 in human liver tissue sections.
Affinity Binding Assay Analysis: A representative lot of this antibody bound SOCS-3 with a KD of 9.2 x 10-7 in an affinity binding assay.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service