Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

ZRB1998

Anti-HDAC6 Antibody, clone 2I3 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

EC:3.5.1.98, JM21, KIAA0901, Protein deacetylase HDAC6, Tubulin-lysine deacetylase HDAC6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2I3, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 131.42 kDa
observed mol wt ~130 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... HDAC6(10013)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2I3 is a ZooMAb® rabbit recombinant monoclonal antibody that detects HDAC6. It targets an epitope within 21 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the N-terminal region of human HDAC6.

Application

Quality Control Testing

Evaluated by Western Blotting in human testis tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected HDAC6 in human testis tissue lysate.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected HDAC6 in human testis tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected HDAC6 in K-562 cells.

Affinity Binding Assay: A representative lot of this antibody bound with HDAC6 peptide at least one thousand-fold (100,000X) higher affinity than with non-specific control peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Histone deacetylase 6 (UniProt: Q9UBN7; EC:3.5.1.98; also known as HD6, Protein deacetylase HDAC6, Tubulin-lysine deacetylase HDAC6) is encoded by the HDAC6 (also known as KIAA0901, JM21) gene (Gene ID: 10013) in human. HDAC6 is a member of the histone deacetylase family, characterized by two catalytic domains that enable its enzymatic activity. It also contains a unique C-terminal region with a ubiquitin-binding domain, which allows it to interact with ubiquitinated proteins, distinguishing it from other HDACs. HDAC6 primarily functions as a deacetylase, removing acetyl groups from lysine residues on histones and non-histone proteins. This activity plays a crucial role in regulating gene expression, protein stability, and cellular signaling pathways. HDAC6 is particularly involved in the deacetylation of tubulin, impacting microtubule dynamics and cellular transport processes. HDAC6 is widely expressed in various tissues, with particularly high levels in the brain, immune cells, and muscle tissues. Its expression can be influenced by cellular stress and inflammatory signals, reflecting its role in stress responses. Alterations in HDAC6 expression and activity have been implicated in several diseases, including cancer, neurodegenerative disorders, and inflammatory conditions. In cancer, HDAC6 can promote tumor progression by deacetylating key proteins involved in cell growth and survival. In neurodegenerative diseases, such as Alzheimer′s, HDAC6 is associated with the clearance of misfolded proteins, suggesting a role in maintaining proteostasis. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service