Skip to Content
MilliporeSigma
All Photos(14)

Key Documents

ZRB1790

Sigma-Aldrich

Anti-PDCD4 Antibody, clone 1E20 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E20, recombinant monoclonal

description

1E20 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 51.73 kDa
observed mol wt ~55 kDa

purified by

using Protein A

species reactivity

mouse, human

species reactivity (predicted by homology)

monkey, canine, feline, rat, bovine, horse, porcine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PDCD4(27250)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E20 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Programmed cell death protein 4 (PDCD4). It targets an epitope within 19 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminal half of human Programmed cell death protein 4 (PDCD4).

Application

Quality Control Testing

Evaluated by Western Blotting in RAW264.7 cell lysate.

Western Blotting Analysis: A 1:3,000 dilution of this antibody detected PDCD4 in RAW264.7 cell lysate.

Tested applications

Western Blotting Analysis: A 1:3,000 dilution from a representative lot detected PDCD4 in human colon tissue lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PDCD4 in MCF-7 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected PDCD4 in human prostate tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound PDCD4 with a KD of 2.5 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Programmed cell death protein 4 (UniProt: Q53EL6; also known as Neoplastic transformation inhibitor protein, Nuclear antigen H731-like, Protein 197/15a, PDCD4) is encoded by the PDCD4 (also known as H731) gene (Gene ID: 27250) in human. PDCD4 is a tumor-suppressor protein that is shown to be upregulated during apoptosis. It affects transcription, translation, and signal transduction to suppress tumor growth. Its levels are upregulated in proliferative cells and lower levels are reported in lung cancer and colon carcinoma. Its levels are also down regulated in transitional cell carcinoma. PDCD4 is reported to shuttle between the nucleus and cytoplasm but is predominantly nuclear under normal growth conditions and when phosphorylated at serine 457. It contains two nuclear localization signal sequences (aa 58-64 and 241-250). PDCD4 can undergo phosphorylation at serine 67 by Ribosomal protein S6 kinase beta-1 (RPS6KB1) in response to mitogens, which promotes its proteasomal degradation. PDCD4 is known to undergo methylation at arginine 110 that enhances its interaction with elF4A. The methylated form of PDCD4 promotes tumor viability during nutrient deprivation, which allows tumors to grow more aggressively. This methylation can reinforce the interaction between the MA3 domains and eIF4A thereby forming a stronger interaction and may also make PDCD4 MA3 domains more accessible for interaction with eIF4A. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Goke, R., et al. (2004). Ann. NY Acad. Sci. 1014; 220-1; Fischer, N., et al. (2012). Biochem. Biophys. Res. Commun. 417(6); 29-34; Fay, MM., et al. (2014). J. Biol. Chem. 289(25); 17541-17552).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service