Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

ZRB1289

Sigma-Aldrich

Anti-AUTS2 Antibody, clone 1D8 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Autism susceptibility gene 2 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 1D8
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 138.98 kDa
observed mol wt ~115 kDa

purified by

using Protein A

species reactivity

rat, human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... AUTS2(26053)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1D8 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Autism susceptibility gene 2 protein (AUTS2).

Immunogen

His-tagged recombinant fragment corresponding to 245 amino acids from the C-terminal half of human Autism susceptibility gene 2 protein (AUTS2).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected AUTS2 in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected AUTS2 in lysate from C6 and HepG2 cells and Human brain tissue and in a lysate overexpressing Myc/DDX-FLAG tagged full-length human AUTS2.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected AUTS2 in PC12 cells.

Affinity Binding Assay:: A representative lot of this antibody bound AUTS2 peptide with a KD of 9.5 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Autism susceptibility gene 2 protein (UniProt: Q8WXX7; also known as AUTS2) is encoded by the AUTS2 (also known as KIAA0442) gene (Gene ID: 26053) in human. AUTS2 is a component of a PRC1-like complex that is strongly expressed in brain, skeletal muscle, and kidney. Its expression is also observed in placenta, lung, and leukocytes. Two major forms of human AUTS2 have been described: full-length (1259 aa) and a truncated C-terminal (711 aa), which is produced from an alternative transcription start site in exon 9. Full-length AUTS2 is localized in both the nucleus and cytoplasm, whereas the truncated AUTS2 isoforms are exclusively nuclear. Cytoplasmic AUTS2 is reported to be involved in cell motility and morphogenesis by regulating reorganization of the actin cytoskeleton. AUTS2 is detected in the cell nuclei of glutamatergic neurons in the cerebral cortex during early developmental stages in mouse and as neural development proceeds, it also appears in the cytoplasm of differentiated neurons, including dendrites, axons, and cell bodies. AUTS2 contains several putative nuclear localization signal sequences and two proline-rich domains. Other predicted protein motifs include a PY (Pro-Tyr) motif (PPPY), eight CAG (His) repeats, several protein kinase phosphorylation sites, and putative SH2- and SH3-binding domains. However, the exact role of these motifs has not been analyzed. AUTS2 activates RAC1 via guanine exchange factors (GEFs) that leads to lamellipodia formation in neuroblastoma cell lines. The proline-rich region is important for the interaction with RAC guanine nucleotide exchange factors and the subsequent activation of RAC1, which then promotes lamellipodia formation. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonal. (Ref.: Biel, A., et al. (2022). Front. Mol. Neurosci. 15; 858582; Hori, K., et al. (2022). Cells. 11(1); 11); Hori, K., et al. (2014). Cell Rep. 9(6); 2166-2179).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service