Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

WH0029979M1

Sigma-Aldrich

Monoclonal Anti-UBQLN1 antibody produced in mouse

clone 1C6-A3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DA41, Anti-DSK2, Anti-FLJ90054, Anti-PLIC1, Anti-XDRP1, Anti-ubiquilin 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C6-A3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBQLN1(29979)

Related Categories

General description

This gene encodes an ubiquitin-like protein (ubiquilin) that shares a high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain an N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus are thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This ubiquilin has also been shown to modulate accumulation of presenilin proteins, and it is found in lesions associated with Alzheimer′s and Parkinson′s disease. Two transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

UBQLN1 (AAH39294, 1 a.a. ~ 589 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAESGESGGPPGSQDSAAGAEGAGTPAAAASAEPKIMKVTVKTPKEKEEFAVPENSSVQQFKEEISKRFKSHTDQLVLIFAGKILKDQDTLSQHGIHDGLTVHLVIKTQNRPQDHSAQQTNTAGSNVTTSSTPNSNSTSGSATSNPFGLGGLGGLAGLSSLGLNTTNFSELQSQMQRQLLSNPEMMVQIMENPFVQSMLSNHDLMRQLIMANPQMQQLIQRNPEISHMLNNPDIMRQTLELARNPAMMQEMMRNQDRALSNLESIPGGYNALRRMYTDIQEPMLSAAQEQFGGNPFASLVSNTSSGEGSQPSRTENRDPLPNPWAPQTSQSSSASSGTASTVGGTTGSTASGTSGQSTTAPNLVPGVGASMFNTPGMQSLLQQITENPQLMQNMLSAPYMRSMMQSLSQNPDLAAQMMLNNPLFAGNPQLQEQMRQQLPTFLQQMQNPDTLSAMSNPRAMQALLQIQQGLQTLATEAPGLIPGFTPGLGALGSTGGSSGTNGSNATPSENTSPTAGTTEPGHQQFIQQMLQALAGVNPQLQNPEVRFQQQLEQLSAMGFLNREANLQALIATGGDINAAIERLLGSQPS

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service