Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

WH0006182M1

Sigma-Aldrich

Monoclonal Anti-MRPL12 antibody produced in mouse

clone 3B12-1A3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-5c52, Anti-L12mt, Anti-MGC8610, Anti-MRPL31/34, Anti-MRPL7, Anti-MRPL7/L12, Anti-RPML12, Anti-mitochondrial ribosomal protein L12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3B12-1A3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MRPL12(6182)

Immunogen

MRPL12 (AAH02344, 1 a.a. ~ 198 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MLPAAARPLWGPCLGLRAAAFRLARRQVPCVCAVRHMRSSGHQRCEALAGAPLDNAPKEYPPKIQQLVQDIASLTLLEISDLNELLKKTLKIQDVGLVPMGGVMSGAVPAAAAQEAVEEDIPIAKERTHFTVRLTEAKPVDKVKLIKEIKNYIQGINLVQAKKLVESLPQEIKANVAKAEAEKIKAALEAVGGTVVLE

Biochem/physiol Actions

MRPL12 (mitochondrial ribosomal protein) is involved in the mitochondrial translation. It directly interacts with the mitochondrial RNA polymerase (POLRMT) and triggers the mitochondrial transcriptional process. Research shows that the interaction between MRPL12 and POLRMT indicates its participation in the regulatory mechanism that coordinates mitochondrial transcription with translation and/or ribosome biogenesis during human mitochondrial gene expression. Study shows that MRPL12 may be involved in the differentiation of colonic epithelial cells and its reduced mitochondrial activity have been shown to be linked with tumor formation. It has been reported that mutated expression of MRPL12 associates with the growth retardation, neurological deterioration and mitochondrial translation deficiency.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nina Kočevar et al.
World journal of gastroenterology, 18(11), 1216-1228 (2012-04-03)
To search for and validate differentially expressed proteins in patients with gastric adenocarcinoma. We used two-dimensional gel electrophoresis and mass spectrometry to search for differentially expressed proteins in patients with gastric adenocarcinoma. A set of proteins was validated with immunoblotting.
Valérie Serre et al.
Biochimica et biophysica acta, 1832(8), 1304-1312 (2013-04-23)
Multiple respiratory chain deficiencies represent a common cause of mitochondrial diseases and are associated with a wide range of clinical symptoms. We report a subject, born to consanguineous parents, with growth retardation and neurological deterioration. Multiple respiratory chain deficiency was
L Marty et al.
Genomics, 41(3), 453-457 (1997-05-01)
Mitochondrial activity requires the expression of nuclear genes, whose products are part of multiproteic complexes leading to ATP production and delivery. We recently characterized a growth-activated mRNA encoding the human mitochondrial ribosomal MRPL12 protein, which is thought to act as
Zhibo Wang et al.
The Journal of biological chemistry, 282(17), 12610-12618 (2007-03-06)
The core human mitochondrial transcription machinery comprises a single subunit bacteriophage-related RNA polymerase, POLRMT, the high mobility group box DNA-binding protein h-mtTFA/TFAM, and two transcriptional co-activator proteins, h-mtTFB1 and h-mtTFB2 that also have rRNA methyltransferase activity. Recapitulation of specific initiation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service