Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0005433M1

Sigma-Aldrich

Monoclonal Anti-POLR2D antibody produced in mouse

clone 1E4-A5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-HSRBP4, Anti-HSRPB4, Anti-RBP4, Anti-polymerase (RNA) II (DNA directed) polypeptide D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E4-A5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... POLR2D(5433)

General description

This gene encodes the fourth largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. In yeast, this polymerase subunit is associated with the polymerase under suboptimal growth conditions and may have a stress protective role. A sequence for a ribosomal pseudogene is contained within the 3′ untranslated region of the transcript from this gene. (provided by RefSeq)

Immunogen

POLR2D (AAH17205, 1 a.a. ~ 142 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAAGGSDPRAGDVEEDASQLIFPKGFETAETLLNSEVHMLLEHRKQQNESAEDEQELSEVFMKTLNYTARFSRFKNRETIASVRSLLLQKKLHKFELACLANLCPETAEESKALIPSLEGRFEDEELQQILDDIQTKRSFQY

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proteomics analysis for finding serum markers of ovarian cancer.
Cheng Y, Liu C, Zhang N, et al.
BioMed Research International, 2014, 179040-179040 (2014)
Peiwu Huang et al.
Toxicology letters, 229(1), 101-110 (2014-06-25)
Trichloroethylene (TCE) is an industrial solvent with widespread occupational exposure and also a major environmental contaminant. Occupational medicamentosa-like dermatitis induced by trichloroethylene (OMLDT) is an autoimmune disease and it has become one major hazard in China. In this study, sera

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service