Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WH0005395M1

Sigma-Aldrich

Monoclonal Anti-PMS2 antibody produced in mouse

clone 4G4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-HNPCC4, Anti-PMS2 postmeiotic segregation increased 2 (S. cerevisiae), Anti-PMS2CL, Anti-PMSL2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PMS2(5395)

General description

This gene is one of the PMS2 gene family members found in clusters on chromosome 7. The product of this gene is involved in DNA mismatch repair. It forms a heterodimer with MLH1 and this complex interacts with other complexes bound to mismatched bases. Mutations in this gene are associated with hereditary nonpolyposis colorectal cancer, Turcot syndrome, and are a cause of supratentorial primitive neuroectodermal tumors. Alternatively spliced transcript variants have been observed for this gene. (provided by RefSeq)

Immunogen

PMS2 (NP_000526, 763 a.a. ~ 862 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ERAKLISLPTSKNWTFGPQDVDELIFMLSDSPGVMCRPSRVKQMFASRACRKSVMIGTALNTSEMKKLITHMGEMDHPWNCPHGRPTMRHIANLGVISQN

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shilpa V et al.
Genomics, 104(4), 257-263 (2014-09-07)
The role of defective mismatch repair (MMR) system in ovarian carcinoma is not well defined. The purpose of the study was to determine the relationship between microsatellite instability (MSI), promoter methylation and protein expression of MMR genes in epithelial ovarian
Anne-Sophie Bats et al.
Anticancer research, 34(6), 3211-3215 (2014-06-13)
To assess the feasibility of Microsatellite Instability (MSI) analysis in uterine cavity washings for detecting endometrial cancer in Lynch syndrome. This was a proof-of-concept study in Lynch syndrome patients, scheduled for hysterectomy. At the beginning of surgical procedure, uterine cavity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service