Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5372

Sigma-Aldrich

TPX2, His tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

C20orf1, C20orf2, DIL-2, DIL2, FLS353, GD:C20orf1, HCA519, HCTP4, REPP86, p100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~110 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TPX2(22974)

General description

TPX2 is a nuclear proliferation-associated protein whose expression is restricted to cell cycle phases S, G2, and M. It mainly expressed in the lung carcinoma cell lines and also found in adult placenta, skeletal muscle, thymus, testis, and small intestine and in fetal brain, liver, and kidney.TPX2 is required for targeting STK6 to the spindle apparatus, and that STK6 may regulate the function of TPX2 during spindle assembly. TPX2 is mailnly function in mitotic spindle assembly likely contributes to its role in chromosome stability control and tumor suppression.

Physical form

Supplied in 50 mM sodium phosphate, pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service