Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5310

Sigma-Aldrich

c-KIT (Y823D), active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CD119, PBT, SCFR

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

0.8-1.2 nmol/min·mg

mol wt

~73 kDa

technique(s)

cell culture | stem cell: suitable

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KIT(3815)

General description

c-KIT (Y823D) encodes the ligand-binding chain (alpha) of the gamma interferon receptor which is a heterodimer of IFNGR1 and IFNGR2. c-KIT (Y823D) is a regulator of hematopoietic stem cells during homeostasis and under conditions of infectious stress. A genetic variation in c-KIT (Y823D) is associated with susceptibility to Helicobacter pylori infection. In addition, defects in c-KIT (Y823D) are a cause of mendelian susceptibility to mycobacterial disease, also known as familial disseminated atypical mycobacterial infection. c-KIT (Y823D is also associated with the clinical outcome of HBV infection in Chinese adults.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service