Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5309

Sigma-Aldrich

c-KIT (T670I), active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CD118, PBT, SCFR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

11.2-16.8 nmol/min·mg

mol wt

~73 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KIT(3815)

General description

c-KIT (T670I) is a protein that belongs to the type I cytokine receptor family which combines with a high-affinity converter subunit, gp130, to form a receptor complex that mediates the action of the leukemia inhibitory factor, a polyfunctional cytokine that is involved in cellular differentiation, proliferation and survival in the adult and the embryo. c-KIT (T670I) is a good candidate gene and the mutation of this gene cause Schwartz-Jampel syndrome type 2, a disease belonging to the group of the bent-bone dysplasias. The gene encoding this protein is localized on human chromosome 4q12.

Biochem/physiol Actions

The protein has a role in melanocyte development.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The human tyrosine kinase Kit and its gatekeeper mutant T670I, show different kinetic properties: Implications for drug design.
Kissova M
Bioorganic & Medicinal Chemistry (2016)
Phase II Trial of Nilotinib in Patients With Metastatic Malignant Melanoma Harboring KIT Gene Aberration: A Multicenter Trial of Korean Cancer Study Group (UN10-06).
Lee SJ
Oncologist (2015)
Clinicopathological significance of c-KIT mutation in gastrointestinal stromal tumors: a systematic review and meta-analysis
Lin Yan
Scientific Reports (2015)
A YAC Contig Spanning a Cluster of Human Type III Receptor Protein Tyrosine Kinase Genes (PDGFRA-KIT-KDR) in Chromosome Segment 4q12
R.A.Spritz
Genomics (1994)
Simultaneous Activation of Signals Through gp130, c-kit, and Interleukin-3
Receptor Promotes a Trilineage Blood Cell Production in the Absence of
Terminally Acting Lineage-Specific Factors
Takafumi Kimura
Blood (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service